missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dihydroxynaphthalene 95.0+%, TCI America™

Catalog No. D09565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09565G 5 g
D095625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D09565G Supplier TCI America Supplier No. D09565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 581-43-1
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00004082
InChI Key MNZMMCVIXORAQL-UHFFFAOYSA-N
Synonym 2,6-dihydroxynaphthalene, 2,6-naphthalenediol, 2,6-naphthohydroquinone, 2-hydroxy-6-naphthol, unii-4xx2nd0257, 2,6-dihydroxy naphthalene, 2,6-dihdroxynaphthalene, 2,6-dihydroxy-naphthalene, acmc-209m2x, aronis24267
PubChem CID 93552
IUPAC Name naphthalene-2,6-diol
SMILES C1=CC2=C(C=CC(=C2)O)C=C1O

Specifications

Melting Point 222°C
Quantity 5 g
Formula Weight 160.17
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dihydroxynaphthalene
EINECSNumber (4)-0356
TSCA Yes