missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dihydroxynaphthalene, 98%

CAS: 581-43-1 | C10H8O2 | 160.172 g/mol

Supplier:  Thermo Scientific Chemicals B2146522

 View more versions of this product

Catalog No. AAB2146522


Only null left
Explore available promotions

Description

Description

2,6-Dihydroxynaphthalene is used in the preparation of 1,5-dichloro-2,6-diethynylnaphthalenes. It is used to produce convulsions in mice. It exhibits antiproliferative activity towards certain cells. It is also employed in the preparation of the first-generation rotaxane dendrimer. Further, it is used in the production of several dyes, pigments, fluorescent whiteners, tanning agents, antioxidants and antiseptics. In addition to this, it is converted into amines, esters, ethers and carboxylic derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dihydroxynaphthalene is used in the preparation of 1,5-dichloro-2,6-diethynylnaphthalenes. It is used to produce convulsions in mice. It exhibits antiproliferative activity towards certain cells. It is also employed in the preparation of the first-generation rotaxane dendrimer. Further, it is used in the production of several dyes, pigments, fluorescent whiteners, tanning agents, antioxidants and antiseptics. In addition to this, it is converted into amines, esters, ethers and carboxylic derivatives.

Solubility
Soluble in methanol and hot water. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

2,6-Dihydroxynaphthalene
223°C to 225°C
MFCD00004082
1238082
Soluble in methanol and hot water. Slightly soluble in water.
C1=CC2=C(C=CC(=C2)O)C=C1O
160.172
160.17
581-43-1
C10H8O2
100 g
2,6-dihydroxynaphthalene, 2,6-naphthalenediol, 2,6-naphthohydroquinone, 2-hydroxy-6-naphthol, unii-4xx2nd0257, 2,6-dihydroxy naphthalene, 2,6-dihdroxynaphthalene, 2,6-dihydroxy-naphthalene, acmc-209m2x, aronis24267
MNZMMCVIXORAQL-UHFFFAOYSA-N
naphthalene-2,6-diol
93552
98%

Chemical Identifiers

581-43-1
160.172
MNZMMCVIXORAQL-UHFFFAOYSA-N
93552
C1=CC2=C(C=CC(=C2)O)C=C1O
C10H8O2
MFCD00004082
2,6-dihydroxynaphthalene, 2,6-naphthalenediol, 2,6-naphthohydroquinone, 2-hydroxy-6-naphthol, unii-4xx2nd0257, 2,6-dihydroxy naphthalene, 2,6-dihdroxynaphthalene, 2,6-dihydroxy-naphthalene, acmc-209m2x, aronis24267
naphthalene-2,6-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-465-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only