missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dihydroxybenzoic Acid 98.0+%, TCI America™
$105.89 - $1108.90
Chemical Identifiers
| CAS | 303-07-1 |
|---|---|
| Molecular Formula | C7H6O4 |
| Molecular Weight (g/mol) | 154.12 |
| MDL Number | MFCD00002462 |
| InChI Key | AKEUNCKRJATALU-UHFFFAOYSA-N |
| Synonym | gamma-resorcylic acid, 2-carboxyresorcinol, 2,6-resorcylic acid, 6-hydroxysalicylic acid, benzoic acid, 2,6-dihydroxy, .gamma.-resorcylic acid, unii-rsa5g6vrpv, rsa5g6vrpv, 2,6-dihydroxy benzoic acid, 2,6-dihydroxy-benzoic acid |
| PubChem CID | 9338 |
| ChEBI | CHEBI:68465 |
| IUPAC Name | 2,6-dihydroxybenzoic acid |
| SMILES | OC(=O)C1=C(O)C=CC=C1O |
Chemical Identifiers
| 303-07-1 | |
| 154.12 | |
| AKEUNCKRJATALU-UHFFFAOYSA-N | |
| 9338 | |
| 2,6-dihydroxybenzoic acid |
| C7H6O4 | |
| MFCD00002462 | |
| gamma-resorcylic acid, 2-carboxyresorcinol, 2,6-resorcylic acid, 6-hydroxysalicylic acid, benzoic acid, 2,6-dihydroxy, .gamma.-resorcylic acid, unii-rsa5g6vrpv, rsa5g6vrpv, 2,6-dihydroxy benzoic acid, 2,6-dihydroxy-benzoic acid | |
| CHEBI:68465 | |
| OC(=O)C1=C(O)C=CC=C1O |
Specifications
| 303-07-1 | |
| C7H6O4 | |
| 25 g | |
| AKEUNCKRJATALU-UHFFFAOYSA-N | |
| 2,6-dihydroxybenzoic acid | |
| 9338 | |
| 154.12 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00002462 | |
| gamma-resorcylic acid, 2-carboxyresorcinol, 2,6-resorcylic acid, 6-hydroxysalicylic acid, benzoic acid, 2,6-dihydroxy, .gamma.-resorcylic acid, unii-rsa5g6vrpv, rsa5g6vrpv, 2,6-dihydroxy benzoic acid, 2,6-dihydroxy-benzoic acid | |
| OC(=O)C1=C(O)C=CC=C1O | |
| 154.12 | |
| CHEBI:68465 | |
| ≥98.0% (GC,T) | |
| 2,6-Dihydroxybenzoic Acid |
Safety and Handling
EINECSNumber : (3)-1616
RTECSNumber : DG8578000
TSCA : Yes