Learn More
2,6-Dihydroxybenzoic acid, 98%
CAS: 303-07-1 | C7H6O4 | 154.12 g/mol
$89.15 - $847.34
Chemical Identifiers
| CAS | 303-07-1 |
|---|---|
| Molecular Formula | C7H6O4 |
| Molecular Weight (g/mol) | 154.12 |
| MDL Number | MFCD00002462 |
| InChI Key | AKEUNCKRJATALU-UHFFFAOYSA-N |
| Synonym | gamma-resorcylic acid, 2-carboxyresorcinol, 2,6-resorcylic acid, 6-hydroxysalicylic acid, benzoic acid, 2,6-dihydroxy, .gamma.-resorcylic acid, unii-rsa5g6vrpv, rsa5g6vrpv, 2,6-dihydroxy benzoic acid, 2,6-dihydroxy-benzoic acid |
| PubChem CID | 9338 |
| ChEBI | CHEBI:68465 |
| IUPAC Name | 2,6-dihydroxybenzoic acid |
| SMILES | OC(=O)C1=C(O)C=CC=C1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1448514
|
Thermo Scientific Chemicals
A1448514 |
25 g |
Each for $89.15
|
|
|||||
|
AAA1448522
|
Thermo Scientific Chemicals
A1448522 |
100 g |
Each for $236.55
|
|
|||||
|
AAA1448536
|
Thermo Scientific Chemicals
A1448536 |
500 g |
Each for $847.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 303-07-1 | |
| 154.12 | |
| AKEUNCKRJATALU-UHFFFAOYSA-N | |
| 9338 | |
| 2,6-dihydroxybenzoic acid |
| C7H6O4 | |
| MFCD00002462 | |
| gamma-resorcylic acid, 2-carboxyresorcinol, 2,6-resorcylic acid, 6-hydroxysalicylic acid, benzoic acid, 2,6-dihydroxy, .gamma.-resorcylic acid, unii-rsa5g6vrpv, rsa5g6vrpv, 2,6-dihydroxy benzoic acid, 2,6-dihydroxy-benzoic acid | |
| CHEBI:68465 | |
| OC(=O)C1=C(O)C=CC=C1O |
Specifications
| 303-07-1 | |
| C7H6O4 | |
| 25 g | |
| gamma-resorcylic acid, 2-carboxyresorcinol, 2,6-resorcylic acid, 6-hydroxysalicylic acid, benzoic acid, 2,6-dihydroxy, .gamma.-resorcylic acid, unii-rsa5g6vrpv, rsa5g6vrpv, 2,6-dihydroxy benzoic acid, 2,6-dihydroxy-benzoic acid | |
| OC(=O)C1=C(O)C=CC=C1O | |
| 154.12 | |
| CHEBI:68465 | |
| 98% |
| ∼164°C (decomposition) | |
| MFCD00002462 | |
| 2209755 | |
| AKEUNCKRJATALU-UHFFFAOYSA-N | |
| 2,6-dihydroxybenzoic acid | |
| 9338 | |
| 154.12 | |
| 2,6-Dihydroxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
EINECSNumber : 206-134-8
RTECSNumber : DG8578000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only