missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dihydroxyacetophenone, Matrix Substance for MALDI-MS, ≥99.5%, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 699-83-2 |
| Linear Formula | (HO)2C6H3 COCH3 |
| MDL Number | MFCD00002270 |
| Melting Point | 156°C to 158°C |
| Synonym | 2-Acetyl-1,3-dihydroxybenzene; 2-Acetylresorcinol; DHAP |
| Solubility | Soluble in dioxane |
| Type | Matrix Substance for MALDI-MS |
| Quantity | 5 g |
| Formula Weight | 152.15 |
| Percent Purity | ≥99.5% (HPLC); ≥99.5% |
| Show More |