missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dihydroxyacetophenone, Matrix Substance for MALDI-MS, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111015280
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015280 1 g
111015281 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015280 Supplier Merck Emd Millipore Supplier No. 374681GF
Only null left

Specifications

CAS 699-83-2
Linear Formula (HO)2C6H3 COCH3
MDL Number MFCD00002270
Melting Point 156°C to 158°C
Synonym 2-Acetyl-1,3-dihydroxybenzene; 2-Acetylresorcinol; DHAP
Solubility Soluble in dioxane
Type Matrix Substance for MALDI-MS
Quantity 1 g
Formula Weight 152.15
Percent Purity ≥99.5% (HPLC); ≥99.5%
Grade Matrix Substance for MALDI-MS
Show More Show Less