missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorotoluene 99.0+%, TCI America™

Catalog No. D23385G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23385G 5 g
D23381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23385G Supplier TCI America Supplier No. D23385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 443-84-5
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.122
MDL Number MFCD00043898
InChI Key MZLSNIREOQCDED-UHFFFAOYSA-N
Synonym 2,6-difluorotoluene, benzene, 1,3-difluoro-2-methyl, 2.6-difluoro methyl benzene, 1,3-difluoro-2-methyl-benzene, 1,3-difluor-2-methylbenzol, pubchem1614, 2,6-difluoro toluene, acmc-209jxo, ksc493c9b
PubChem CID 581493
IUPAC Name 1,3-difluoro-2-methylbenzene
SMILES CC1=C(C=CC=C1F)F

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 g
UN Number 1993
Formula Weight 128.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Difluorotoluene