missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluorophenylacetone, 98%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | (F)2C6H3CH2COCH3 |
| Quantity | 1g |
| Formula Weight | 170.16 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,6-Difluorophenylacetone |
RUO – Research Use Only