missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorophenyl isocyanate, 97%

Catalog No. p-7046530
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1477903 1 g
AAL1477906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1477903 Supplier Thermo Scientific Chemicals Supplier No. L1477903

CAS: 65295-69-4 | C7H3F2NO | 155.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65295-69-4
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.104
MDL Number MFCD00001998
InChI Key YXHDLKWTPVMIOH-UHFFFAOYSA-N
Synonym 2,6-difluorophenyl isocyanate, 2,6-difluorophenylisocyanate, benzene, 1,3-difluoro-2-isocyanato, 2,6-difluorobenzenisocyanate, timtec-bb sbb006664, pubchem5023, acmc-1bcwu, 2,6 difluorophenylisocyanate, 2,6-difluoro phenyl isocyanate, 1-isocyanato-2,6-difluoro benzene
PubChem CID 522162
IUPAC Name 1,3-difluoro-2-isocyanatobenzene
SMILES C1=CC(=C(C(=C1)F)N=C=O)F

Specifications

Melting Point 22°C to 25°C
Boiling Point 60°C (10 mmHg)
Flash Point 58°C (136°F)
Refractive Index 1.4928
Quantity 1 g
UN Number UN2930
Beilstein 7115873
Sensitivity Moisture sensitive
Formula Weight 155.11
Percent Purity 97%
Chemical Name or Material 2,6-Difluorophenyl isocyanate
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H300-H330-H334-H228-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only