missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluorophenyl isocyanate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 22°C to 25°C |
| Boiling Point | 60°C (10 mmHg) |
| Flash Point | 58°C (136°F) |
| Refractive Index | 1.4928 |
| Quantity | 5 g |
| UN Number | UN2930 |
| Beilstein | 7115873 |
| Sensitivity | Moisture sensitive |
| Formula Weight | 155.11 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H226-H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H300-H330-H334-H228-H315-H319-H335 Fatal if swallowed. Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only