missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluorobenzyl Chloride 98.0+%, TCI America™
$57.75 - $225.25
Chemical Identifiers
| CAS | 697-73-4 |
|---|---|
| Molecular Formula | C7H5ClF2 |
| Molecular Weight (g/mol) | 162.564 |
| MDL Number | MFCD00191346 |
| InChI Key | MJXRENZUAQXZGJ-UHFFFAOYSA-N |
| PubChem CID | 581570 |
| IUPAC Name | 2-(chloromethyl)-1,3-difluorobenzene |
| SMILES | C1=CC(=C(C(=C1)F)CCl)F |
Chemical Identifiers
| 697-73-4 | |
| 162.564 | |
| MJXRENZUAQXZGJ-UHFFFAOYSA-N | |
| 2-(chloromethyl)-1,3-difluorobenzene |
| C7H5ClF2 | |
| MFCD00191346 | |
| 581570 | |
| C1=CC(=C(C(=C1)F)CCl)F |
Specifications
| 697-73-4 | |
| MFCD00191346 | |
| 3261 | |
| MJXRENZUAQXZGJ-UHFFFAOYSA-N | |
| 2-(chloromethyl)-1,3-difluorobenzene | |
| 581570 | |
| ≥98.0% (GC) | |
| 2,6-Difluorobenzyl Chloride |
| C7H5ClF2 | |
| 5 g | |
| 2,6-difluorobenzyl chloride, 2-chloromethyl-1,3-difluorobenzene, 2,6-difluorobenzylchloride, benzene, 2-chloromethyl-1,3-difluoro, alpha-chloro-2,6-difluorotoluene, 2, 6-difluorobenzyl chloride, 1-chloromethyl-2,6-difluorobenzene, 2-chlormethyl-1,3-difluorbenzol, pubchem2178 | |
| C1=CC(=C(C(=C1)F)CCl)F | |
| 162.564 | |
| 162.56 | |
| Crystalline Powder |
Safety and Handling
TSCA : No