missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzyl bromide, 96%

Catalog No. AAA1743414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1743414 25 g
AAA1743406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1743414 Supplier Thermo Scientific Chemicals Supplier No. A1743414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85118-00-9 | C7H5BrF2 | 207.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85118-00-9
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.02
MDL Number MFCD00000329
InChI Key LSXJPJGBWSZHTM-UHFFFAOYSA-N
Synonym 2,6-difluorobenzyl bromide, 2-bromomethyl-1,3-difluorobenzene, 2,6-difluorobenzylbromide, alpha-bromo-2,6-difluorotoluene, benzene, 2-bromomethyl-1,3-difluoro, timtec-bb sbb006564, 2-bromomethyl-1,3-difluoro-benzene, fr cf b1e, pubchem4915, acmc-209q4j
PubChem CID 581435
IUPAC Name 2-(bromomethyl)-1,3-difluorobenzene
SMILES FC1=CC=CC(F)=C1CBr

Specifications

Melting Point 49°C to 53°C
Density 1.6
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN1759
Beilstein 2083943
Formula Weight 207.02
Percent Purity ≥97%
Chemical Name or Material 2,6-Difluorobenzyl bromide
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 285-652-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only