missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluorobenzyl Alcohol 97.0+%, TCI America™
$82.02 - $238.60
Chemical Identifiers
| CAS | 19064-18-7 |
|---|---|
| Molecular Formula | C7H6F2O |
| Molecular Weight (g/mol) | 144.121 |
| MDL Number | MFCD00004603 |
| InChI Key | LVICICZQETYOGS-UHFFFAOYSA-N |
| Synonym | 2,6-difluorobenzyl alcohol, 2,6-difluorophenyl methanol, benzenemethanol, 2,6-difluoro, 2,6-difluorobenzylalcohol, 2,6-difluorobenzenemethanol, 2,6-difluoro-phenyl-methanol, 2,6-difluoro benzyl alcohol, 2,6-difluorophenyl methan-1-ol, pubchem4914, acmc-209etz |
| PubChem CID | 87921 |
| IUPAC Name | (2,6-difluorophenyl)methanol |
| SMILES | C1=CC(=C(C(=C1)F)CO)F |
Chemical Identifiers
| 19064-18-7 | |
| 144.121 | |
| LVICICZQETYOGS-UHFFFAOYSA-N | |
| 87921 | |
| C1=CC(=C(C(=C1)F)CO)F |
| C7H6F2O | |
| MFCD00004603 | |
| 2,6-difluorobenzyl alcohol, 2,6-difluorophenyl methanol, benzenemethanol, 2,6-difluoro, 2,6-difluorobenzylalcohol, 2,6-difluorobenzenemethanol, 2,6-difluoro-phenyl-methanol, 2,6-difluoro benzyl alcohol, 2,6-difluorophenyl methan-1-ol, pubchem4914, acmc-209etz | |
| (2,6-difluorophenyl)methanol |
Specifications
| 19064-18-7 | |
| 88°C | |
| MFCD00004603 | |
| 2,6-difluorobenzyl alcohol, 2,6-difluorophenyl methanol, benzenemethanol, 2,6-difluoro, 2,6-difluorobenzylalcohol, 2,6-difluorobenzenemethanol, 2,6-difluoro-phenyl-methanol, 2,6-difluoro benzyl alcohol, 2,6-difluorophenyl methan-1-ol, pubchem4914, acmc-209etz | |
| C1=CC(=C(C(=C1)F)CO)F | |
| 144.121 | |
| 144.12 | |
| Liquid |
| Yellow | |
| C7H6F2O | |
| 5 g | |
| LVICICZQETYOGS-UHFFFAOYSA-N | |
| (2,6-difluorophenyl)methanol | |
| 87921 | |
| ≥97.0% (GC) | |
| 2,6-Difluorobenzyl Alcohol |
Safety and Handling
EINECSNumber : (3)-4498
TSCA : No