missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzoyl Chloride 97.0+%, TCI America™

Catalog No. d279825g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D279825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D279825G Supplier TCI America Supplier No. D279825G
Only null left

Chemical Identifiers

CAS 18063-02-0
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00000659
InChI Key QRHUZEVERIHEPT-UHFFFAOYSA-N
Synonym 2,6-difluorobenzoylchloride, benzoyl chloride, 2,6-difluoro, 2,6-difluoro-benzoyl chloride, acmc-1ca8w, 2,6-diflurobenzoyl chloride, 2 6-difluorobenzoyl chloride, 2,6-diflourobenzoyl chloride, 2,6-difluoro benzoylchloride, 2.6-difluorobenzoyl chloride, 2,6-difluro-benzoyl chloride
PubChem CID 87438
IUPAC Name 2,6-difluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)F)C(=O)Cl)F

Specifications

Color Yellow
Quantity 25 g
UN Number 2920
Formula Weight 176.55
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,6-Difluorobenzoyl Chloride