missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzoic Acid 98.0+%, TCI America™

Catalog No. D162925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D162925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D162925G Supplier TCI America Supplier No. D162925G

Chemical Identifiers

CAS 385-00-2
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00002411
InChI Key ONOTYLMNTZNAQZ-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-difluoro, 2,6-difluorobenzoicacid, 2,6-difluoro benzoic acid, 2,6-difluoro-benzoic acid, pubchem2274, benzoic acid,6-difluoro, acmc-209j0h, 2,6,-difluorobenzoic acid, 3-09-00-01330 beilstein handbook reference, ksc223c7l
PubChem CID 9796
IUPAC Name 2,6-difluorobenzoic acid
SMILES C1=CC(=C(C(=C1)F)C(=O)O)F

Specifications

Melting Point 158°C
Boiling Point 77°C
Quantity 25 g
Formula Weight 158.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluorobenzoic Acid
RTECSNumber DG8559000
TSCA No