missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzoic acid, 98+%

Catalog No. p-7021992
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1455814 25 g
AAA1455822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1455814 Supplier Thermo Scientific Chemicals Supplier No. A1455814

CAS: 385-00-2 | C7H4F2O2 | 158.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 385-00-2
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00002411
InChI Key ONOTYLMNTZNAQZ-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-difluoro, 2,6-difluorobenzoicacid, 2,6-difluoro benzoic acid, 2,6-difluoro-benzoic acid, pubchem2274, benzoic acid,6-difluoro, acmc-209j0h, 2,6,-difluorobenzoic acid, 3-09-00-01330 beilstein handbook reference, ksc223c7l
PubChem CID 9796
IUPAC Name 2,6-difluorobenzoic acid
SMILES C1=CC(=C(C(=C1)F)C(=O)O)F

Specifications

Melting Point 156°C to 160°C
Boiling Point 72°C to 77°C (13 mmHg)
Odor Odorless
Quantity 25 g
Beilstein 973774
Formula Weight 158.11
Percent Purity ≥98%
Chemical Name or Material 2,6-Difluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-856-3
RTECSNumber DG8559000
TSCA Yes

RUO – Research Use Only