missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7127304
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24525G 5 g
D245225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24525G Supplier TCI America Supplier No. D24525G
Only null left

Chemical Identifiers

CAS 437-81-0
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.105
MDL Number MFCD00010293
InChI Key SOWRUJSGHKNOKN-UHFFFAOYSA-N
Synonym 2,6-difluoro benzaldehyde, 2,6-dfad, benzaldehyde, 2,6-difluoro, 2,6-difluorobenzaledehyde, pubchem1438, 2,6-difluorbenzaldehyde, 2,6 difluorobenzaldehyde, acmc-1adi4, 2, 6-difluorobenzaldehyde, 2,6-difluoro-benzaldehyde
PubChem CID 136284
IUPAC Name 2,6-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)F)C=O)F

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
Formula Weight 142.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Difluorobenzaldehyde