missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoroaniline, 98%

Catalog No. AAA1302022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1302022 100 g
AAA1302006 5 g
AAA1302014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1302022 Supplier Thermo Scientific Chemicals Supplier No. A1302022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5509-65-9 | C6H5F2N | 129.11 g/mol

2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acid chlorides, acid anhydrides, oxidizing agents.

Chemical Identifiers

CAS 5509-65-9
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007655
InChI Key ODUZJBKKYBQIBX-UHFFFAOYSA-N
Synonym benzenamine, 2,6-difluoro, 2,6-difluoro-phenylamine, 2,6-difluoro aniline, 2,6-difluorobenzenamine, 2,6-difluoranilin, 2,6 difluoroaniline, 2,6-diflouroaniline, pubchem4034, 2,6-difuluoroaniline
PubChem CID 79647
IUPAC Name 2,6-difluoroaniline
SMILES NC1=C(F)C=CC=C1F

Specifications

Density 1.275
Boiling Point 152°C to 154°C
Flash Point 51°C (124°F)
Odor Characteristic
Refractive Index 1.508
Quantity 100 g
UN Number UN1992
Beilstein 2802697
Solubility Information Slightly soluble in water.
Formula Weight 129.11
Percent Purity 98%
Chemical Name or Material 2,6-Difluoroaniline
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H331-H226-H373-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 226-853-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only