missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-4-iodophenol, 99%

Catalog No. AAH3208009
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3208009 10 g
AAH3208003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3208009 Supplier Thermo Scientific Chemicals Supplier No. H3208009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 950858-06-7 | C6H3F2IO | 255.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.

Chemical Identifiers

CAS 950858-06-7
Molecular Formula C6H3F2IO
Molecular Weight (g/mol) 255.99
MDL Number MFCD09907951
InChI Key LHUADYKKTXZZMQ-UHFFFAOYSA-N
PubChem CID 45790937
IUPAC Name 2,6-difluoro-4-iodophenol
SMILES C1=C(C=C(C(=C1F)O)F)I

Specifications

Melting Point 70°C to 73°C
Quantity 10 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 255.99
Percent Purity 99%
Chemical Name or Material 2,6-Difluoro-4-iodophenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only