missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluoro-4-hydroxybenzoic Acid 98.0+%, TCI America™
$95.06 - $419.41
Chemical Identifiers
| CAS | 214917-68-7 |
|---|---|
| Molecular Formula | C7H4F2O3 |
| Molecular Weight (g/mol) | 174.10 |
| MDL Number | MFCD03094500 |
| InChI Key | NFIQGYBXSJQLSR-UHFFFAOYSA-N |
| Synonym | 2,6-difluoro-4-hydroxy-benzoic acid, benzoic acid, 2,6-difluoro-4-hydroxy, pubchem4938, acmc-1cles, ksc495o6f, 4-carboxy-3,5-difluorophenol |
| PubChem CID | 2778776 |
| IUPAC Name | 2,6-difluoro-4-hydroxybenzoic acid |
| SMILES | OC(=O)C1=C(F)C=C(O)C=C1F |
Chemical Identifiers
| 214917-68-7 | |
| 174.10 | |
| NFIQGYBXSJQLSR-UHFFFAOYSA-N | |
| 2778776 | |
| OC(=O)C1=C(F)C=C(O)C=C1F |
| C7H4F2O3 | |
| MFCD03094500 | |
| 2,6-difluoro-4-hydroxy-benzoic acid, benzoic acid, 2,6-difluoro-4-hydroxy, pubchem4938, acmc-1cles, ksc495o6f, 4-carboxy-3,5-difluorophenol | |
| 2,6-difluoro-4-hydroxybenzoic acid |
Specifications
| 214917-68-7 | |
| White-Yellow | |
| MFCD03094500 | |
| 2,6-difluoro-4-hydroxy-benzoic acid, benzoic acid, 2,6-difluoro-4-hydroxy, pubchem4938, acmc-1cles, ksc495o6f, 4-carboxy-3,5-difluorophenol | |
| OC(=O)C1=C(F)C=C(O)C=C1F | |
| 174.10 | |
| 174.10 | |
| Crystalline Powder |
| 176°C | |
| C7H4F2O3 | |
| 1 g | |
| NFIQGYBXSJQLSR-UHFFFAOYSA-N | |
| 2,6-difluoro-4-hydroxybenzoic acid | |
| 2778776 | |
| ≥98.0% (GC) | |
| 2,6-Difluoro-4-hydroxybenzoic Acid |
Safety and Handling
TSCA : No