missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-4-hydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. D39851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39851G 1 g
D39855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39851G Supplier TCI America Supplier No. D39851G

Chemical Identifiers

CAS 214917-68-7
Molecular Formula C7H4F2O3
Molecular Weight (g/mol) 174.10
MDL Number MFCD03094500
InChI Key NFIQGYBXSJQLSR-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-hydroxy-benzoic acid, benzoic acid, 2,6-difluoro-4-hydroxy, pubchem4938, acmc-1cles, ksc495o6f, 4-carboxy-3,5-difluorophenol
PubChem CID 2778776
IUPAC Name 2,6-difluoro-4-hydroxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(O)C=C1F

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 1 g
Formula Weight 174.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluoro-4-hydroxybenzoic Acid