missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-4-hydroxybenzoic acid, 95%

Catalog No. AAH26134MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26134MD 250 mg
AAH2613403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26134MD Supplier Thermo Scientific Chemicals Supplier No. H26134MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 214917-68-7 | C7H4F2O3 | 174.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 214917-68-7
Molecular Formula C7H4F2O3
Molecular Weight (g/mol) 174.10
MDL Number MFCD03094500
InChI Key NFIQGYBXSJQLSR-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-hydroxy-benzoic acid, benzoic acid, 2,6-difluoro-4-hydroxy, pubchem4938, acmc-1cles, ksc495o6f, 4-carboxy-3,5-difluorophenol
PubChem CID 2778776
IUPAC Name 2,6-difluoro-4-hydroxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(O)C=C1F

Specifications

Melting Point 170°C to 171°C (decomposition)
Quantity 250 mg
Formula Weight 174.1
Percent Purity 95%
Chemical Name or Material 2,6-Difluoro-4-hydroxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only