missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
$57.79 - $169.63
Chemical Identifiers
| CAS | 1072945-00-6 |
|---|---|
| Molecular Formula | C11H14BF2NO2 |
| Molecular Weight (g/mol) | 241.04 |
| MDL Number | MFCD09037475 |
| InChI Key | CLDFZDCZPCIBKO-UHFFFAOYSA-N |
| Synonym | 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-difluoropyridine-3-boronic acid pinacol ester, 2,6-difluoropyridin-3-ylboronic acid pinacol ester, 2,6-difluoropyridin-3-ylboronic acid, pinacol ester, 2,6-difluoropyridin-3-yl boronic acid pinacol ester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl pyridine, 2,6-difluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-2,6-difluoro-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| PubChem CID | 44755153 |
| IUPAC Name | 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
| SMILES | CC1(C)OB(OC1(C)C)C1=C(F)N=C(F)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D5142200MG
|
TCI America
D5142200MG |
200 mg |
Each for $57.79
|
|
|||||
|
D51421G
|
TCI America
D51421G |
1 g |
Each for $169.63
|
|
|||||
Chemical Identifiers
| 1072945-00-6 | |
| 241.04 | |
| CLDFZDCZPCIBKO-UHFFFAOYSA-N | |
| 44755153 | |
| CC1(C)OB(OC1(C)C)C1=C(F)N=C(F)C=C1 |
| C11H14BF2NO2 | |
| MFCD09037475 | |
| 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-difluoropyridine-3-boronic acid pinacol ester, 2,6-difluoropyridin-3-ylboronic acid pinacol ester, 2,6-difluoropyridin-3-ylboronic acid, pinacol ester, 2,6-difluoropyridin-3-yl boronic acid pinacol ester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl pyridine, 2,6-difluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-2,6-difluoro-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Specifications
| 1072945-00-6 | |
| White | |
| MFCD09037475 | |
| 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-difluoropyridine-3-boronic acid pinacol ester, 2,6-difluoropyridin-3-ylboronic acid pinacol ester, 2,6-difluoropyridin-3-ylboronic acid, pinacol ester, 2,6-difluoropyridin-3-yl boronic acid pinacol ester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl pyridine, 2,6-difluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-2,6-difluoro-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| CC1(C)OB(OC1(C)C)C1=C(F)N=C(F)C=C1 | |
| 241.04 | |
| 241.04 | |
| Crystalline Powder |
| 57°C | |
| C11H14BF2NO2 | |
| 200 mg | |
| CLDFZDCZPCIBKO-UHFFFAOYSA-N | |
| 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
| 44755153 | |
| ≥98.0% (GC,T) | |
| 2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Safety and Handling
TSCA : No