accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNFF

CAS RN 1072945-00-6

IUPAC Name: 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 241.04
Molecular Formula: C11H14BF2NO2
InChi Key: CLDFZDCZPCIBKO-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)N=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results