missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. D5142200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5142200MG 200 mg
D51421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5142200MG Supplier TCI America Supplier No. D5142200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1072945-00-6
Molecular Formula C11H14BF2NO2
Molecular Weight (g/mol) 241.04
MDL Number MFCD09037475
InChI Key CLDFZDCZPCIBKO-UHFFFAOYSA-N
Synonym 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-difluoropyridine-3-boronic acid pinacol ester, 2,6-difluoropyridin-3-ylboronic acid pinacol ester, 2,6-difluoropyridin-3-ylboronic acid, pinacol ester, 2,6-difluoropyridin-3-yl boronic acid pinacol ester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl pyridine, 2,6-difluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-2,6-difluoro-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 44755153
IUPAC Name 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=C(F)N=C(F)C=C1

Specifications

Melting Point 57°C
Color White
Quantity 200 mg
Formula Weight 241.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine