missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorotoluene, 99+%

Catalog No. AC113791000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113791000 100 mL Glass bottle
AC113795000 500 mL Glass bottle
AC113790025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC113791000 Supplier Thermo Scientific Chemicals Supplier No. 113791000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 118-69-4 | C7H6Cl2 | 161.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-69-4
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.03
MDL Number MFCD00000576
InChI Key DMEDNTFWIHCBRK-UHFFFAOYSA-N
Synonym 2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene
PubChem CID 8368
IUPAC Name 1,3-dichloro-2-methylbenzene
SMILES CC1=C(C=CC=C1Cl)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 2°C
Color Colorless to Yellow
Density 1.2500g/mL
Boiling Point 196°C to 203°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5500 to 1.552
Linear Formula CH3C6H3Cl2
Quantity 100 mL
Beilstein 05, 296
Specific Gravity 1.25
Solubility Information Solubility in water: 24mg/l (20°C). Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 161.03
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2, 6-Dichlorotoluene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 204-269-7
TSCA TSCA

RUO – Research Use Only