Learn More
2,6-Dichlorotoluene, 99%
CAS: 118-69-4 | C7H6Cl2 | 161.025 g/mol
Supplier: Thermo Scientific Chemicals B2446322
Description
2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
| 2,6-Dichlorotoluene | |
| 2°C | |
| 196°C to 203°C | |
| C7H6Cl2 | |
| MFCD00000576 | |
| UN3082 | |
| 2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene | |
| DMEDNTFWIHCBRK-UHFFFAOYSA-N | |
| 1,3-dichloro-2-methylbenzene | |
| 8368 | |
| 99% |
| 118-69-4 | |
| 1.254 | |
| 82°C (179°F) | |
| 1.551 | |
| 100 g | |
| 1907194 | |
| Soluble in water. | |
| CC1=C(C=CC=C1Cl)Cl | |
| 161.025 | |
| 161.03 |
Chemical Identifiers
| 118-69-4 | |
| 161.025 | |
| DMEDNTFWIHCBRK-UHFFFAOYSA-N | |
| 8368 | |
| CC1=C(C=CC=C1Cl)Cl |
| C7H6Cl2 | |
| MFCD00000576 | |
| 2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene | |
| 1,3-dichloro-2-methylbenzene |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 204-269-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only