missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorothiophenol, 97%

Catalog No. AAA1285406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1285406 5 g
AAA1285414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1285406 Supplier Thermo Scientific Chemicals Supplier No. A1285406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24966-39-0 | C6H3Cl2S | 178.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24966-39-0
Molecular Formula C6H3Cl2S
Molecular Weight (g/mol) 178.05
MDL Number MFCD00004833
InChI Key JBISHCXLCGVPGW-UHFFFAOYSA-M
Synonym 2,6-dichlorothiophenol, 2,6-dichlorobenzene-1-thiol, 2,6-dichloro thiophenol, benzenethiol, 2,6-dichloro, pubchem6802, 2,6-dichloro-benzenethiol, acmc-209gg5, benzenethiol,2,6-dichloro
PubChem CID 90671
IUPAC Name 2,6-dichlorobenzenethiol
SMILES [S-]C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 46°C to 50°C
Boiling Point 101°C to 103°C (14 mmHg)
Flash Point 112°C (233°F)
Odor Unpleasant
Quantity 5 g
Beilstein 2082400
Sensitivity Air Sensitive
Formula Weight 179.07
Percent Purity 97%
Chemical Name or Material 2,6-Dichlorothiophenol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 416-720-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only