missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorostyrene, 96%, stab. with 0.1% 4-tert-butylcatechol

Catalog No. AAL0786006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0786006 5 g
AAL0786014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0786006 Supplier Thermo Scientific Chemicals Supplier No. L0786006

CAS: 28469-92-3 | C8H6Cl2 | 173.04 g/mol

2,6-Dichlorostyrene is used in the preparation of 1,3-dichloro-2-(1,2-dibromo-ethyl)-benzene by bromination reaction using bromine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichlorostyrene is used in the preparation of 1,3-dichloro-2-(1,2-dibromo-ethyl)-benzene by bromination reaction using bromine.

Solubility
Immiscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 28469-92-3
Molecular Formula C8H6Cl2
Molecular Weight (g/mol) 173.04
MDL Number MFCD00000579
InChI Key YJCVRMIJBXTMNR-UHFFFAOYSA-N
Synonym 2,6-dichlorostyrene, 1,3-dichloro-2-vinylbenzene, benzene, 1,3-dichloro-2-ethenyl, 2,6-dichlorstyrol, acmc-20ao92, 1,3-dichloro-2-vinylbenzene #, 2,6-dichlorostyrene, stabilized 5g
PubChem CID 34254
IUPAC Name 1,3-dichloro-2-ethenylbenzene
SMILES ClC1=CC=CC(Cl)=C1C=C

Specifications

Density 1.266
Boiling Point 88°C to 90°C (8 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.574
Quantity 5 g
Beilstein 1931527
Solubility Information Immiscible with water.
Formula Weight 173.04
Percent Purity 96%
Chemical Name or Material 2,6-Dichlorostyrene, stabilized with 0.1% 4-tert-butylcatechol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 249-039-7
RTECSNumber CZ4907000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only