Learn More
2,6-Dichloropyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals
$248.90 - $248.90
Chemical Identifiers
| CAS | 5398-44-7 |
|---|---|
| Molecular Formula | C6H3Cl2NO2 |
| Molecular Weight (g/mol) | 192.00 |
| MDL Number | MFCD00234147 |
| InChI Key | SQSYNRCXIZHKAI-UHFFFAOYSA-N |
| Synonym | 2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403 |
| PubChem CID | 94830 |
| ChEBI | CHEBI:73179 |
| IUPAC Name | 2,6-dichloropyridine-4-carboxylic acid |
| SMILES | OC(=O)C1=CC(Cl)=NC(Cl)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2054106
|
Thermo Scientific Chemicals
B2054106 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5398-44-7 | |
| 192.00 | |
| SQSYNRCXIZHKAI-UHFFFAOYSA-N | |
| 94830 | |
| 2,6-dichloropyridine-4-carboxylic acid |
| C6H3Cl2NO2 | |
| MFCD00234147 | |
| 2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403 | |
| CHEBI:73179 | |
| OC(=O)C1=CC(Cl)=NC(Cl)=C1 |
Specifications
| 5398-44-7 | |
| C6H3Cl2NO2 | |
| 5 g | |
| SQSYNRCXIZHKAI-UHFFFAOYSA-N | |
| 2,6-dichloropyridine-4-carboxylic acid | |
| 94830 | |
| 192 | |
| 2,6-Dichloropyridine-4-carboxylic acid |
| 208°C (decomposition) | |
| MFCD00234147 | |
| 2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403 | |
| OC(=O)C1=CC(Cl)=NC(Cl)=C1 | |
| 192.00 | |
| CHEBI:73179 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only