missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2054106

 View more versions of this product

Catalog No. AAB2054106

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,6-Dichloropyridine-4-carboxylic acid
208°C (decomposition)
MFCD00234147
2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403
OC(=O)C1=CC(Cl)=NC(Cl)=C1
192.00
CHEBI:73179
98%
5398-44-7
C6H3Cl2NO2
5 g
SQSYNRCXIZHKAI-UHFFFAOYSA-N
2,6-dichloropyridine-4-carboxylic acid
94830
192

Chemical Identifiers

5398-44-7
192.00
SQSYNRCXIZHKAI-UHFFFAOYSA-N
94830
2,6-dichloropyridine-4-carboxylic acid
C6H3Cl2NO2
MFCD00234147
2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403
CHEBI:73179
OC(=O)C1=CC(Cl)=NC(Cl)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only