missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropurine 97.0+%, TCI America™

Catalog No. p-7127315
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24701G 1 g
D24705G 5 g
D247025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D24701G Supplier TCI America Supplier No. D24701G
Only null left

Chemical Identifiers

CAS 5451-40-1
Molecular Formula C5H2Cl2N4
Molecular Weight (g/mol) 189.00
MDL Number MFCD00077725
InChI Key RMFWVOLULURGJI-UHFFFAOYSA-N
Synonym 2,6-dichloropurine, 2,6-dichloro-9h-purine, 2,6-dichloro-1h-purine, 9h-purine, 2,6-dichloro, 2,6-dichloro purine, 2,6 dichloropurine, 2,6-dichioropurine, purine,6-dichloro, zlchem 226, 2,6-dichloro-purine
PubChem CID 5324412
IUPAC Name 2,6-dichloro-7H-purine
SMILES ClC1=NC(Cl)=C2NC=NC2=N1

Specifications

Quantity 1 g
Formula Weight 189.00
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloropurine