missing translation for 'onlineSavingsMsg'
Learn More

(2,6-Dichlorophenylthio)acetic acid, 99%

Catalog No. AAH5028203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5028203 1 g
AAH5028206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5028203 Supplier Thermo Scientific Chemicals Supplier No. H5028203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21248-45-3 | C8H5Cl2O2S | 236.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21248-45-3
Molecular Formula C8H5Cl2O2S
Molecular Weight (g/mol) 236.09
MDL Number MFCD00052108
InChI Key URNRRSJGOOTQBO-UHFFFAOYSA-M
Synonym 2,6-dichlorophenylthio acetic acid, 2-2,6-dichlorophenyl thio acetic acid, 2,6-dichlorophenyl thio acetic acid, 2,6-dichlorophenyl sulfanyl acetic acid, 2-2,6-dichlorophenyl sulfanylacetic acid, acmc-1cg2g, 2,6-dichlorothiophenoxyacetic acid, 2-2,6-dichlorophenylthio acetic acid, 2-2,6-dichlorophenyl sulfanyl acetic acid
PubChem CID 1810591
IUPAC Name 2-(2,6-dichlorophenyl)sulfanylacetic acid
SMILES [O-]C(=O)CSC1=C(Cl)C=CC=C1Cl

Specifications

Quantity 1 g
Formula Weight 237.11
Percent Purity 99%
Chemical Name or Material (2,6-Dichlorophenylthio)acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only