missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylhydrazine hydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB08663EA

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. BTB08663EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

50709-36-9
213.486
2,6-dichlorophenylhydrazine hydrochloride, 2,6-dichlorophenyl hydrazine hydrochloride, 1-2,6-dichlorophenyl hydrazine hydrochloride, 2,6-dichlorophenyl hydrazine monohydrochloride, hydrazine, 2,6-dichlorophenyl-, monohydrochloride, 2,6-dichlorophenylhydrazinehydrochloride, acmc-209kpe, ksc491e8j, 2,6-dichlorophenylhydrazine, chloride, 2,6-dichlorophenylhydrazine hcl
(2,6-dichlorophenyl)hydrazine;hydrochloride
C6H7Cl3N2
CQNIYLLTIOPFCJ-UHFFFAOYSA-N
2723909
C1=CC(=C(C(=C1)Cl)NN)Cl.Cl

Specifications

1-(2, 6-Dichlorophenyl)hydrazine hydrochloride
97%
C6H7Cl3N2
10g
CQNIYLLTIOPFCJ-UHFFFAOYSA-N
(2,6-dichlorophenyl)hydrazine;hydrochloride
2723909
97%
50709-36-9
Amber Glass Bottle
Cl2C6H3NHNH2·HCl
2,6-dichlorophenylhydrazine hydrochloride, 2,6-dichlorophenyl hydrazine hydrochloride, 1-2,6-dichlorophenyl hydrazine hydrochloride, 2,6-dichlorophenyl hydrazine monohydrochloride, hydrazine, 2,6-dichlorophenyl-, monohydrochloride, 2,6-dichlorophenylhydrazinehydrochloride, acmc-209kpe, ksc491e8j, 2,6-dichlorophenylhydrazine, chloride, 2,6-dichlorophenylhydrazine hcl
C1=CC(=C(C(=C1)Cl)NN)Cl.Cl
213.486
213.49
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents