missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylhydrazine hydrochloride, 97%, Thermo Scientific™

Catalog No. BTB08663DA
Change view
Click to view available options
Quantity:
1g
10g
25g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB08663DA 1g Amber Glass Bottle
BTB08663EA 10g Amber Glass Bottle
BTB08663EB 25g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB08663DA Supplier Thermo Scientific Chemicals Supplier No. BTB08663DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50709-36-9
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
InChI Key CQNIYLLTIOPFCJ-UHFFFAOYSA-N
Synonym 2,6-dichlorophenylhydrazine hydrochloride, 2,6-dichlorophenyl hydrazine hydrochloride, 1-2,6-dichlorophenyl hydrazine hydrochloride, 2,6-dichlorophenyl hydrazine monohydrochloride, hydrazine, 2,6-dichlorophenyl-, monohydrochloride, 2,6-dichlorophenylhydrazinehydrochloride, acmc-209kpe, ksc491e8j, 2,6-dichlorophenylhydrazine, chloride, 2,6-dichlorophenylhydrazine hcl
PubChem CID 2723909
IUPAC Name (2,6-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C(=C1)Cl)NN)Cl.Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3NHNH2·HCl
Quantity 1g
Formula Weight 213.49
Percent Purity 97%
Chemical Name or Material 1-(2, 6-Dichlorophenyl)hydrazine hydrochloride