missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylacetic acid, 98%

CAS: 6575-24-2 | C8H6Cl2O2 | 205.034 g/mol

$49.42 - $393.66

Chemical Identifiers

CAS 6575-24-2
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00004320
InChI Key SFAILOOQFZNOAU-UHFFFAOYSA-N
Synonym 2,6-dichlorophenylacetic acid, 2-2,6-dichlorophenyl acetic acid, 2,6-dichlorophenyl acetic acid, benzeneacetic acid, 2,6-dichloro, 2,6-dichloro benzeneacetic acid, 2,6-dichlorophenylaceticacid, unii-k8g025cx8w, 2,6-dichlorobenzeneaceticacid, 2cl
PubChem CID 81058
IUPAC Name 2-(2,6-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)CC(=O)O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2135406
View Documents
Thermo Scientific Chemicals
B2135406
5 g
Each for $49.42
Only null left
 
AAB2135414
View Documents
Thermo Scientific Chemicals
B2135414
25 g
Each for $129.40
Only null left
 
AAB2135422
View Documents
Thermo Scientific Chemicals
B2135422
100 g
Each for $393.66
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6575-24-2
205.034
SFAILOOQFZNOAU-UHFFFAOYSA-N
81058
C1=CC(=C(C(=C1)Cl)CC(=O)O)Cl
C8H6Cl2O2
MFCD00004320
2,6-dichlorophenylacetic acid, 2-2,6-dichlorophenyl acetic acid, 2,6-dichlorophenyl acetic acid, benzeneacetic acid, 2,6-dichloro, 2,6-dichloro benzeneacetic acid, 2,6-dichlorophenylaceticacid, unii-k8g025cx8w, 2,6-dichlorobenzeneaceticacid, 2cl
2-(2,6-dichlorophenyl)acetic acid

Specifications

6575-24-2
C8H6Cl2O2
5 g
2,6-dichlorophenylacetic acid, 2-2,6-dichlorophenyl acetic acid, 2,6-dichlorophenyl acetic acid, benzeneacetic acid, 2,6-dichloro, 2,6-dichloro benzeneacetic acid, 2,6-dichlorophenylaceticacid, unii-k8g025cx8w, 2,6-dichlorobenzeneaceticacid, 2cl
C1=CC(=C(C(=C1)Cl)CC(=O)O)Cl
205.034
205.04
2,6-Dichlorophenylacetic acid
158°C to 161°C
MFCD00004320
1952744
SFAILOOQFZNOAU-UHFFFAOYSA-N
2-(2,6-dichlorophenyl)acetic acid
81058
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 229-504-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only