missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenyl isothiocyanate, 98+%

Catalog No. p-7045980
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1267203 1 g
AAL1267206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1267203 Supplier Thermo Scientific Chemicals Supplier No. L1267203

CAS: 6590-95-0 | C7H3Cl2NS | 204.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6590-95-0
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00041056
InChI Key SUCGVQHNGIQXGD-UHFFFAOYSA-N
Synonym 2,6-dichlorophenyl isothiocyanate, 2,6-dichlorophenylisothiocyanate, benzene, 1,3-dichloro-2-isothiocyanato, 1,3-dichloro-2-isothiocyanato-benzene, benzene,1,3-dichloro-2-isothiocyanato, c7h3c12ns, 2,6-dichlorobenzenisothiocyanate, 2,6-dichlorophenyl-isothiocyanate, 1,3-dichloro-2-thioisocyanato-benzene, 2,6-dichloro-1-isothiocyanato-benzene
PubChem CID 123239
IUPAC Name 1,3-dichloro-2-isothiocyanatobenzene
SMILES ClC1=CC=CC(Cl)=C1N=C=S

Specifications

Melting Point 42°C to 44°C
Boiling Point 122°C to 124°C (8 mmHg)
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 639061
Sensitivity Moisture sensitive
Formula Weight 204.08
Percent Purity ≥98%
Chemical Name or Material 2,6-Dichlorophenyl isothiocyanate
Hazard Category H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only