missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenyl Isocyanate 98.0+%, TCI America™

Catalog No. D36685G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36685G Supplier TCI America Supplier No. D36685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39920-37-1
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00002003
InChI Key HMVKMAMIRAVXAN-UHFFFAOYSA-N
Synonym 2,6-dichlorophenyl isocyanate, benzene, 1,3-dichloro-2-isocyanato, 2,6-dichlorophenylisocyanate, isocyanic acid 2,6-dichlorophenyl ester, pubchem23916, acmc-1ailv, 2,6 dichlorophenylisocyanate, 2,6-dichlorobenzenisocyanate, 2,6dichlorophenyl isocyanate, 1,3-dichloro-2-isocyanato-benzene
PubChem CID 604537
IUPAC Name 1,3-dichloro-2-isocyanatobenzene
SMILES C1=CC(=C(C(=C1)Cl)N=C=O)Cl

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 130°C
Quantity 5 g
UN Number 2250
Formula Weight 188.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorophenyl Isocyanate