accessibility menu, dialog, popup

UserName

CAS RN 39920-37-1

IUPAC Name: 1,3-dichloro-2-isocyanatobenzene
Synonyms: 1,3-dichloro-2-isocyanatobenzene
Molecular Weight (g/mol): 188.01
Molecular Formula: C7H3Cl2NO
InChi Key: HMVKMAMIRAVXAN-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1N=C=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)