Learn More
2,6-Dichlorophenyl isocyanate, 98%
CAS: 39920-37-1 | C7H3Cl2NO | 188.007 g/mol
$163.82 - $163.82
Chemical Identifiers
| CAS | 39920-37-1 |
|---|---|
| Molecular Formula | C7H3Cl2NO |
| Molecular Weight (g/mol) | 188.007 |
| MDL Number | MFCD00002003 |
| InChI Key | HMVKMAMIRAVXAN-UHFFFAOYSA-N |
| Synonym | 2,6-dichlorophenyl isocyanate, benzene, 1,3-dichloro-2-isocyanato, 2,6-dichlorophenylisocyanate, isocyanic acid 2,6-dichlorophenyl ester, pubchem23916, acmc-1ailv, 2,6 dichlorophenylisocyanate, 2,6-dichlorobenzenisocyanate, 2,6dichlorophenyl isocyanate, 1,3-dichloro-2-isocyanato-benzene |
| PubChem CID | 604537 |
| IUPAC Name | 1,3-dichloro-2-isocyanatobenzene |
| SMILES | C1=CC(=C(C(=C1)Cl)N=C=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1042106
|
Thermo Scientific Chemicals
L1042106 |
5 g |
Each for $163.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 39920-37-1 | |
| 188.007 | |
| HMVKMAMIRAVXAN-UHFFFAOYSA-N | |
| 604537 | |
| C1=CC(=C(C(=C1)Cl)N=C=O)Cl |
| C7H3Cl2NO | |
| MFCD00002003 | |
| 2,6-dichlorophenyl isocyanate, benzene, 1,3-dichloro-2-isocyanato, 2,6-dichlorophenylisocyanate, isocyanic acid 2,6-dichlorophenyl ester, pubchem23916, acmc-1ailv, 2,6 dichlorophenylisocyanate, 2,6-dichlorobenzenisocyanate, 2,6dichlorophenyl isocyanate, 1,3-dichloro-2-isocyanato-benzene | |
| 1,3-dichloro-2-isocyanatobenzene |
Specifications
| 39920-37-1 | |
| 101°C (5 mmHg) | |
| C7H3Cl2NO | |
| 5 g | |
| 608323 | |
| 2,6-dichlorophenyl isocyanate, benzene, 1,3-dichloro-2-isocyanato, 2,6-dichlorophenylisocyanate, isocyanic acid 2,6-dichlorophenyl ester, pubchem23916, acmc-1ailv, 2,6 dichlorophenylisocyanate, 2,6-dichlorobenzenisocyanate, 2,6dichlorophenyl isocyanate, 1,3-dichloro-2-isocyanato-benzene | |
| C1=CC(=C(C(=C1)Cl)N=C=O)Cl | |
| 188.007 | |
| 188.01 | |
| 2,6-Dichlorophenyl isocyanate |
| 42°C to 46°C | |
| 76°C (168°F) | |
| MFCD00002003 | |
| UN2250 | |
| Moisture sensitive | |
| HMVKMAMIRAVXAN-UHFFFAOYSA-N | |
| 1,3-dichloro-2-isocyanatobenzene | |
| 604537 | |
| 98% |
Safety and Handling
GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P363-P501c
H301+H311+H331-H315-H319-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROPHENYL ISOCYANATES
EINECSNumber : 254-699-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only