missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophénol 99,0+%, TCI America™

Numéro de catalogue. D039425G
Modifier l'affichage
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D039425G 25 g
D0394500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D039425G Fournisseur TCI America Code fournisseur D039425G

Identifiants chimiques

CAS 87-65-0
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002176
InChI Key HOLHYSJJBXSLMV-UHFFFAOYSA-N
Synonym phenol, 2,6-dichloro, 2,6-dichlorfenol, 2,6-dichlorphenol, rcra waste number u082, 2,6-dichloro-phenol, 2,6-dichlorfenol czech, unii-q7e9k52w7e, 2,6 dichlorophenol, ccris 2511, 2,6-dcp
PubChem CID 6899
ChEBI CHEBI:28457
IUPAC Name 2,6-dichlorophénol
SMILES OC1=C(Cl)C=CC=C1Cl

Spécifications

Melting Point 66°C
Color Blanc-jaune
Boiling Point 80°C
Quantity 25 g
UN Number 2020
Formula Weight 163.00
Percent Purity ≥99.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 2,6-Dichlorophenol
EINECSNumber (3)-0930
RTECSNumber SK8750000
TSCA Yes