missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenol, SPEX CertiPrep™

Catalog No. S1415
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S1415 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S1415 Supplier SPEX CertiPrep Group LLC Supplier No. S1415

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2,6-Dichlorophenol87-65-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 87-65-0 , 67-56-1
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002176
InChI Key HOLHYSJJBXSLMV-UHFFFAOYSA-N
PubChem CID 6899
ChEBI CHEBI:28457
IUPAC Name 2,6-dichlorophenol
SMILES OC1=C(Cl)C=CC=C1Cl

Specifications

Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methanol
Quantity 1 mL
Chemical Name or Material 2,6-Dichlorophenol
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only