missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenol, 99%

CAS: 87-65-0 | C6H4Cl2O | 163.00 g/mol

Supplier:  Thermo Scientific Chemicals 113590010

 View more versions of this product

Catalog No. AC113590010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 6-Dichlorophenol
87-65-0
100.0
White to Yellow
Authentic
Glass bottle
Cl2C6H3OH
1 kg
15, 3083
Solubility in water: 1.9g/L (25°C). Other solubilities: soluble in petroleum ether,benzene,diethyl ether,and ethanol
OC1=C(Cl)C=CC=C1Cl
163.00
CHEBI:28457
99%
99%
98.5
65.0°C to 68.0°C
218.0°C to 220.0°C
98.5% min. (GC)
C6H4Cl2O
MFCD00002176
06, 190
phenol, 2,6-dichloro, 2,6-dichlorfenol, 2,6-dichlorphenol, rcra waste number u082, 2,6-dichloro-phenol, 2,6-dichlorfenol czech, unii-q7e9k52w7e, 2,6 dichlorophenol, ccris 2511, 2,6-dcp
HOLHYSJJBXSLMV-UHFFFAOYSA-N
2,6-dichlorophenol
6899
163
Crystalline Powder or Crystals

Chemical Identifiers

87-65-0
163.00
HOLHYSJJBXSLMV-UHFFFAOYSA-N
6899
2,6-dichlorophenol
C6H4Cl2O
MFCD00002176
phenol, 2,6-dichloro, 2,6-dichlorfenol, 2,6-dichlorphenol, rcra waste number u082, 2,6-dichloro-phenol, 2,6-dichlorfenol czech, unii-q7e9k52w7e, 2,6 dichlorophenol, ccris 2511, 2,6-dcp
CHEBI:28457
OC1=C(Cl)C=CC=C1Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 201-761-3

RTECSNumber : SK8750000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only