missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenol, 99%

Catalog No. AAA1441136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1441136 500 g
AAA1441122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1441136 Supplier Thermo Scientific Chemicals Supplier No. A1441136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-65-0 | C6H4Cl2O | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-65-0
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002176
InChI Key HOLHYSJJBXSLMV-UHFFFAOYSA-N
Synonym phenol, 2,6-dichloro, 2,6-dichlorfenol, 2,6-dichlorphenol, rcra waste number u082, 2,6-dichloro-phenol, 2,6-dichlorfenol czech, unii-q7e9k52w7e, 2,6 dichlorophenol, ccris 2511, 2,6-dcp
PubChem CID 6899
ChEBI CHEBI:28457
IUPAC Name 2,6-dichlorophenol
SMILES OC1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 65°C to 68°C
Boiling Point 218°C to 220°C
Odor Phenol-like
Quantity 500 g
UN Number UN2020
Beilstein 1447806
Merck Index 14,3073
Formula Weight 163
Percent Purity 99%
Chemical Name or Material 2,6-Dichlorophenol
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
EINECSNumber 201-761-3
RTECSNumber SK8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only