missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorodiphenylamine, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111020074
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020074 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020074 Supplier Merck Emd Millipore Supplier No. 75538100MG

Specifications

CAS 15307-93-4
Flash Point 110°C - Closed Cup
Linear Formula Cl2C6H3 NHC6H5
MDL Number MFCD00269648
Melting Point 50°C to 53°C
Synonym N-Phenyl-2,6-dichloroaniline
Type Analytical Standard
Quantity 100 mg
Formula Weight 238.11
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less