missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorocinnamic Acid 98.0+%, TCI America™
$50.33 - $195.73
Chemical Identifiers
| CAS | 5345-89-1 |
|---|---|
| Molecular Formula | C9H6Cl2O2 |
| Molecular Weight (g/mol) | 217.05 |
| MDL Number | MFCD00004374 |
| InChI Key | OIPVGRCXMFBNAN-SNAWJCMRSA-N |
| Synonym | 2,6-dichlorocinnamic acid, 3-2,6-dichlorophenyl acrylic acid, e-3-2,6-dichlorophenyl acrylic acid, 2e-3-2,6-dichlorophenyl prop-2-enoic acid, 2e-3-2,6-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,6-dichlorophenyl, 3-2,6-dichlorophenyl-2-propenoic acid, cinnamic acid, 2,6-dichloro-6ci,7ci, 3-2,6-dichlorophenyl propenoic acid, 2,6-dichlorocinnamic acid, predominantly trans |
| PubChem CID | 731762 |
| IUPAC Name | (2E)-3-(2,6-dichlorophenyl)prop-2-enoic acid |
| SMILES | OC(=O)\C=C\C1=C(Cl)C=CC=C1Cl |
Chemical Identifiers
| 5345-89-1 | |
| 217.05 | |
| OIPVGRCXMFBNAN-SNAWJCMRSA-N | |
| 731762 | |
| OC(=O)\C=C\C1=C(Cl)C=CC=C1Cl |
| C9H6Cl2O2 | |
| MFCD00004374 | |
| 2,6-dichlorocinnamic acid, 3-2,6-dichlorophenyl acrylic acid, e-3-2,6-dichlorophenyl acrylic acid, 2e-3-2,6-dichlorophenyl prop-2-enoic acid, 2e-3-2,6-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,6-dichlorophenyl, 3-2,6-dichlorophenyl-2-propenoic acid, cinnamic acid, 2,6-dichloro-6ci,7ci, 3-2,6-dichlorophenyl propenoic acid, 2,6-dichlorocinnamic acid, predominantly trans | |
| (2E)-3-(2,6-dichlorophenyl)prop-2-enoic acid |
Specifications
| 5345-89-1 | |
| White-Yellow | |
| MFCD00004374 | |
| 2,6-dichlorocinnamic acid, 3-2,6-dichlorophenyl acrylic acid, e-3-2,6-dichlorophenyl acrylic acid, 2e-3-2,6-dichlorophenyl prop-2-enoic acid, 2e-3-2,6-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,6-dichlorophenyl, 3-2,6-dichlorophenyl-2-propenoic acid, cinnamic acid, 2,6-dichloro-6ci,7ci, 3-2,6-dichlorophenyl propenoic acid, 2,6-dichlorocinnamic acid, predominantly trans | |
| OC(=O)\C=C\C1=C(Cl)C=CC=C1Cl | |
| 217.05 | |
| 217.05 | |
| Crystalline Powder |
| 196°C | |
| C9H6Cl2O2 | |
| 5 g | |
| OIPVGRCXMFBNAN-SNAWJCMRSA-N | |
| (2E)-3-(2,6-dichlorophenyl)prop-2-enoic acid | |
| 731762 | |
| ≥98.0% (GC,T) | |
| 2,6-Dichlorocinnamic Acid |
Safety and Handling
RTECSNumber : UD3333800
TSCA : No