CAS RN 5345-89-1
IUPAC Name:
(2E)-3-(2,6-dichlorophenyl)prop-2-enoic acid
Synonyms:
(2E)-3-(2,6-dichlorophenyl)prop-2-enoic acid
Molecular Weight (g/mol):
217.05
Molecular Formula:
C9H6Cl2O2
InChi Key:
OIPVGRCXMFBNAN-SNAWJCMRSA-N
SMILES:
OC(=O)\C=C\C1=C(Cl)C=CC=C1Cl
1
–
1
of
1
results
1
2,6-Dichlorocinnamic Acid 98.0+%, TCI America™
CAS: 5345-89-1 Molecular Formula: C9H6Cl2O2 Molecular Weight (g/mol): 217.05 MDL Number: MFCD00004374 InChI Key: OIPVGRCXMFBNAN-SNAWJCMRSA-N Synonym: 2,6-dichlorocinnamic acid,3-2,6-dichlorophenyl acrylic acid,e-3-2,6-dichlorophenyl acrylic acid,2e-3-2,6-dichlorophenyl prop-2-enoic acid,2e-3-2,6-dichlorophenyl acrylic acid,2-propenoic acid, 3-2,6-dichlorophenyl,3-2,6-dichlorophenyl-2-propenoic acid,cinnamic acid, 2,6-dichloro-6ci,7ci,3-2,6-dichlorophenyl propenoic acid,2,6-dichlorocinnamic acid, predominantly trans PubChem CID: 731762 IUPAC Name: (2E)-3-(2,6-dichlorophenyl)prop-2-enoic acid SMILES: OC(=O)\C=C\C1=C(Cl)C=CC=C1Cl