missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzylamine 97.0+%, TCI America™

Catalog No. p-7128318
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45215G 5 g
D452125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45215G Supplier TCI America Supplier No. D45215G
Only null left

Chemical Identifiers

CAS 6575-27-5
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00047928
InChI Key VLVLNNQMURDGPM-UHFFFAOYSA-N
PubChem CID 485432
IUPAC Name 1-(2,6-dichlorophenyl)methanamine
SMILES NCC1=C(Cl)C=CC=C1Cl

Specifications

Color Yellow
Boiling Point 244°C
Quantity 5 g
UN Number 2735
Synonym 2,6-dichlorobenzylamine, 2,6-dichlorophenyl methanamine, benzenemethanamine, 2,6-dichloro, 2,6-dichlorophenyl methylamine, 1-2,6-dichlorophenyl methanamine, pubchem15690, 2,6-dichloro-benzylamine, acmc-20ao98, chembl13305, benzenemethanamine,2,6-dichloro
Formula Weight 176.04
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,6-Dichlorobenzylamine