missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzylamine, 98%

Catalog No. p-2689844
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC350740050 5 g Glass bottle
AC350740250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC350740050 Supplier Thermo Scientific Chemicals Supplier No. 350740050

CAS: 6575-27-5 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6575-27-5
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00047928
InChI Key VLVLNNQMURDGPM-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzylamine, 2,6-dichlorophenyl methanamine, benzenemethanamine, 2,6-dichloro, 2,6-dichlorophenyl methylamine, 1-2,6-dichlorophenyl methanamine, pubchem15690, 2,6-dichloro-benzylamine, acmc-20ao98, chembl13305, benzenemethanamine,2,6-dichloro
PubChem CID 485432
IUPAC Name (2,6-dichlorophenyl)methanamine
SMILES NCC1=C(Cl)C=CC=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5570 to 1.559
Quantity 5 g
Beilstein 12, III, 2347
Formula Weight 176.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 6-Dichlorobenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only