missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzylamine, 97+%

Catalog No. AAH2927414
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2927414 25 g
AAH2927403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2927414 Supplier Thermo Scientific Chemicals Supplier No. H2927414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6575-27-5 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6575-27-5
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00047928
InChI Key VLVLNNQMURDGPM-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzylamine, 2,6-dichlorophenyl methanamine, benzenemethanamine, 2,6-dichloro, 2,6-dichlorophenyl methylamine, 1-2,6-dichlorophenyl methanamine, pubchem15690, 2,6-dichloro-benzylamine, acmc-20ao98, chembl13305, benzenemethanamine,2,6-dichloro
PubChem CID 485432
IUPAC Name (2,6-dichlorophenyl)methanamine
SMILES NCC1=C(Cl)C=CC=C1Cl

Specifications

Odor Amine-like
Quantity 25 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 176.05
Percent Purity ≥97%
Chemical Name or Material 2,6-Dichlorobenzylamine
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only